ENA444724848 Enamine

N1-(1-phenylethyl)benzene-1,2-diamine

  • MDL number MFCD11187008
  • Cas Number : 1039987-39-7
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-199817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199817
Availability details for
Quantity
  • Availability
  • Add to Cart