ENA444724866 Enamine

methyl 4,5-dihydro-1,2-oxazole-5-carboxylate

  • MDL number MFCD18833575
  • Cas Number : 15055-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202525
Availability details for
Quantity
  • Availability
  • Add to Cart