ENA444724868 Enamine

(cyclobutylmethyl)[(2,4-dimethoxyphenyl)methyl]amine

  • MDL number MFCD11634814
  • Cas Number : 1096278-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202533
Availability details for
Quantity
  • Availability
  • Add to Cart