ENA444724869 Enamine

3-bromo-2-cyanobenzene-1-sulfonyl chloride

  • MDL number MFCD18390730
  • Cas Number : 612541-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202535
Availability details for
Quantity
  • Availability
  • Add to Cart