ENA444724871 Enamine

3-methylcyclopentane-1-carbaldehyde

  • MDL number MFCD20862544
  • Cas Number : 82879-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202538
Availability details for
Quantity
  • Availability
  • Add to Cart