ENA444724890 Enamine

(1S)-2-amino-1-(3-chlorophenyl)ethan-1-ol hydrochloride

  • MDL number MFCD11042304
  • Cas Number : 469887-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202586
Availability details for
Quantity
  • Availability
  • Add to Cart