ENA444724893 Enamine

(3,3-dimethylcyclobutyl)methanol

  • MDL number MFCD17170398
  • Cas Number : 75017-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202603
Availability details for
Quantity
  • Availability
  • Add to Cart