ENA444724896 Enamine

4-(naphthalen-1-yloxy)benzonitrile

  • MDL number MFCD12737236
  • Cas Number : 1041608-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202700
Availability details for
Quantity
  • Availability
  • Add to Cart