ENA444724897 Enamine

4-(3,5-dichlorophenoxy)benzonitrile

  • MDL number MFCD12563819
  • Cas Number : 1153351-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202701
Availability details for
Quantity
  • Availability
  • Add to Cart