ENA444724898 Enamine

2-(difluoromethoxy)-5-nitropyridine

  • MDL number MFCD11847317
  • Cas Number : 1192813-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202702
Availability details for
Quantity
  • Availability
  • Add to Cart