ENA444724906 Enamine

(3-fluorophenyl)(3-methylphenyl)methanone

  • MDL number MFCD12153249
  • Cas Number : 864087-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202733
Availability details for
Quantity
  • Availability
  • Add to Cart