ENA444724908 Enamine

1-(1,2-dimethyl-1H-imidazol-4-yl)ethan-1-one

  • MDL number MFCD18829313
  • Cas Number : 103747-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202737
Availability details for
Quantity
  • Availability
  • Add to Cart