ENA444724914 Enamine

1-(2-bromo-1-methoxyethyl)-4-chlorobenzene

  • MDL number MFCD16680784
  • Cas Number : 83739-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202748
Availability details for
Quantity
  • Availability
  • Add to Cart