ENA444724918 Enamine

3-carbamoyl-2-methylpropanoic acid

  • MDL number MFCD19205075
  • Cas Number : 98071-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202757
Availability details for
Quantity
  • Availability
  • Add to Cart