ENA444724919 Enamine

(2,6-dibromophenyl)hydrazine

  • MDL number MFCD22565113
  • Cas Number : 14763-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202758
Availability details for
Quantity
  • Availability
  • Add to Cart