ENA444725104 Enamine

3-(3-methyl-1H-pyrazol-5-yl)piperidine hydrochloride

  • MDL number MFCD28348159

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202796
Availability details for
Quantity
  • Availability
  • Add to Cart