ENA444725105 Enamine

1-(4-chloro-2-fluorophenyl)-1-(1H-1,2,3-triazol-4-yl)methanamine hydrochloride

  • MDL number MFCD28348160
  • Cas Number : 1803594-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202806
Availability details for
Quantity
  • Availability
  • Add to Cart