ENA444725108 Enamine

1-[3-(3-fluoro-4-methoxyphenyl)-4,5-dihydro-1,2-oxazol-5-yl]methanamine hydrochloride

  • MDL number MFCD28348163
  • Cas Number : 1187947-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202872
Availability details for
Quantity
  • Availability
  • Add to Cart