ENA444725112 Enamine

2-methylpyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD28348167
  • Cas Number : 1803589-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202928
Availability details for
Quantity
  • Availability
  • Add to Cart