ENA444725113 Enamine

methoxy[(oxan-4-yl)methyl]amine hydrochloride

  • MDL number MFCD28348168
  • Cas Number : 1803585-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202931
Availability details for
Quantity
  • Availability
  • Add to Cart