ENA444725116 Enamine

2,5-diethyl (2R,5S)-pyrrolidine-2,5-dicarboxylate hydrochloride

  • MDL number MFCD28348171
  • Cas Number : 162376-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202935
Availability details for
Quantity
  • Availability
  • Add to Cart