ENA444725119 Enamine

O-(1-methylpiperidin-4-yl)hydroxylamine dihydrochloride

  • MDL number MFCD28348175
  • Cas Number : 132710-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203001
Availability details for
Quantity
  • Availability
  • Add to Cart