ENA444725123 Enamine

N-(2-{[(thiophen-3-yl)methyl]amino}ethyl)acetamide hydrochloride

  • MDL number MFCD28348182
  • Cas Number : 1803596-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203045
Availability details for
Quantity
  • Availability
  • Add to Cart