ENA444725131 Enamine

5-(morpholin-4-yl)-2-(piperidin-3-yl)-2,3-dihydropyridazin-3-one; carbonic acid

  • MDL number MFCD28348193
  • Cas Number : 1803590-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data OC(=O)O
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203126
Availability details for
Quantity
  • Availability
  • Add to Cart