ENA444725134 Enamine

2-chloro-3-nitroaniline hydrochloride

  • MDL number MFCD28348198
  • Cas Number : 1803590-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203201
Availability details for
Quantity
  • Availability
  • Add to Cart