ENA444725137 Enamine

5-(furan-2-yl)-4-methyl-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD28348201
  • Cas Number : 1803600-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203239
Availability details for
Quantity
  • Availability
  • Add to Cart