ENA444725140 Enamine

N-(prop-2-en-1-yl)cyclooctanamine hydrochloride

  • MDL number MFCD28348206
  • Cas Number : 1803603-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203301
Availability details for
Quantity
  • Availability
  • Add to Cart