ENA444725141 Enamine

4-hydroxypiperidine-4-carboxamide hydrochloride

  • MDL number MFCD28348207
  • Cas Number : 240400-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203306
Availability details for
Quantity
  • Availability
  • Add to Cart