ENA444725142 Enamine

5-fluoro-6-methoxy-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD28348208
  • Cas Number : 1803611-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203312
Availability details for
Quantity
  • Availability
  • Add to Cart