ENA444725148 Enamine

methyl 5-methyl-octahydrofuro[3,2-b]pyridine-2-carboxylate hydrochloride

  • MDL number MFCD28348214
  • Cas Number : 1803561-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203388
Availability details for
Quantity
  • Availability
  • Add to Cart