ENA444725153 Enamine

9-(trifluoromethyl)-6-azaspiro[3.5]nonane

  • MDL number MFCD28348221
  • Cas Number : 1803582-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203439
Availability details for
Quantity
  • Availability
  • Add to Cart