ENA444725154 Enamine

6-chloro-4-hydroxy-5-methyl-1,2-dihydropyridin-2-one hydrochloride

  • MDL number MFCD28348224
  • Cas Number : 95306-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203458
Availability details for
Quantity
  • Availability
  • Add to Cart