ENA444725155 Enamine

3-(3,5-dimethoxyphenyl)-3-fluoropyrrolidine hydrochloride

  • MDL number MFCD28348225
  • Cas Number : 1803604-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203459
Availability details for
Quantity
  • Availability
  • Add to Cart