ENA444725156 Enamine

methyl[2-(3-nitrophenyl)ethyl]amine hydrochloride

  • MDL number MFCD28348226
  • Cas Number : 1803604-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203465
Availability details for
Quantity
  • Availability
  • Add to Cart