ENA444725158 Enamine

1-{2-[(1H-1,2,4-triazol-1-yl)methyl]phenyl}methanamine dihydrochloride

  • MDL number MFCD28348228
  • Cas Number : 1803594-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203472

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203472
Availability details for
Quantity
  • Availability
  • Add to Cart