ENA444725161 Enamine

1-(3-aminopiperidin-1-yl)-2-phenylethan-1-one hydrochloride

  • MDL number MFCD28348232
  • Cas Number : 1803586-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203480
Availability details for
Quantity
  • Availability
  • Add to Cart