ENA444725168 Enamine

1-{4,5,7-trimethyl-4H,7H-[1,2,3,4]tetrazolo[1,5-a]pyrimidin-6-yl}ethan-1-one

  • MDL number MFCD28348240
  • Cas Number : 1803611-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203611
Availability details for
Quantity
  • Availability
  • Add to Cart