ENA444725173 Enamine

4-fluoro-4-(3-fluoro-5-methylphenyl)piperidine hydrochloride

  • MDL number MFCD28348245
  • Cas Number : 1803603-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203628
Availability details for
Quantity
  • Availability
  • Add to Cart