ENA444725190 Enamine

[4-(1,3-dioxaindan-5-yl)butan-2-yl]hydrazine dihydrochloride

  • MDL number MFCD28348262
  • Cas Number : 1803586-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203777
Availability details for
Quantity
  • Availability
  • Add to Cart