ENA444725196 Enamine

3-{1-[(tert-butoxy)carbonyl]piperidin-3-yl}-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD28348268
  • Cas Number : 1803611-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203840
Availability details for
Quantity
  • Availability
  • Add to Cart