ENA444725197 Enamine

{4,5-dimethyl-4H,7H-[1,2,3,4]tetrazolo[1,5-a]pyrimidin-6-yl}methanol

  • MDL number MFCD28348270
  • Cas Number : 1803604-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203859
Availability details for
Quantity
  • Availability
  • Add to Cart