ENA444725198 Enamine

3-(dimethylamino)azetidine-3-carbonitrile dihydrochloride

  • MDL number MFCD28348271
  • Cas Number : 1803604-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203874
Availability details for
Quantity
  • Availability
  • Add to Cart