ENA444725199 Enamine

1-(pyridin-3-yl)cyclopentane-1-carboxylic acid hydrochloride

  • MDL number MFCD28348273
  • Cas Number : 1803584-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203880
Availability details for
Quantity
  • Availability
  • Add to Cart