ENA444725204 Enamine

7-bromo-2,3,4,5-tetrahydro-1,4-benzoxazepine hydrochloride

  • MDL number MFCD26389238
  • Cas Number : 740842-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203945
Availability details for
Quantity
  • Availability
  • Add to Cart