ENA444725206 Enamine

2-(bromomethyl)-1-(propan-2-yl)piperidine hydrobromide

  • MDL number MFCD28348279
  • Cas Number : 1803597-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204030
Availability details for
Quantity
  • Availability
  • Add to Cart