ENA444725208 Enamine

1-[4-(3,5-dichlorophenoxy)phenyl]methanamine hydrochloride

  • MDL number MFCD28348281
  • Cas Number : 1803582-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204035
Availability details for
Quantity
  • Availability
  • Add to Cart