ENA444725209 Enamine

1-(cyclopropanesulfonyl)-1,4-diazepane hydrochloride

  • MDL number MFCD28348282
  • Cas Number : 1403952-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204038
Availability details for
Quantity
  • Availability
  • Add to Cart