ENA444725212 Enamine

2-[1-(propan-2-yl)piperidin-3-yl]acetic acid hydrochloride

  • MDL number MFCD28348285
  • Cas Number : 1803611-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204063
Availability details for
Quantity
  • Availability
  • Add to Cart