ENA444725214 Enamine

N-(prop-2-yn-1-yl)piperidine-4-carboxamide hydrochloride

  • MDL number MFCD28348287
  • Cas Number : 1803586-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204107

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204107
Availability details for
Quantity
  • Availability
  • Add to Cart