ENA444725216 Enamine

7-methanesulfonyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrochloride

  • MDL number MFCD28348289
  • Cas Number : 123018-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204117
Availability details for
Quantity
  • Availability
  • Add to Cart